Anti-CDKN2A Rabbit Polyclonal Antibody (Biotin)

Proveedor: Novus Biologicals
NB200-106B
NOVUNB200-106BEA 710 EUR
NOVUNB200-106B
Anti-CDKN2A Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR