Anti-MMP2 Mouse Monoclonal Antibody [clone: MMP2/8B4] (Biotin)

Proveedor: Novus Biologicals
NB200-114B
NOVUNB200-114BEA 792 EUR
NOVUNB200-114B
Anti-MMP2 Mouse Monoclonal Antibody [clone: MMP2/8B4] (Biotin)
Anticuerpos
Mouse Monoclonal MMP2 Antibody (MMP2/8B4) [Biotin]. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 2 ?g/ml

Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR