Anti-MMP2 Mouse Monoclonal Antibody (DyLight® 755) [clone: 8B4]

Proveedor: Novus Biologicals
NB200-114IR
NOVUNB200-114IREA 792 EUR
NOVUNB200-114IR
Anti-MMP2 Mouse Monoclonal Antibody (DyLight® 755) [clone: 8B4]
Anticuerpos
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: monoclonal
Clone: 8B4
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR