Anti-CFTR Mouse Monoclonal Antibody [clone: CF3]

Proveedor: Novus Biologicals
NB300-511
NOVUNB300-511EA 0 EUR
NOVUNB300-511
Anti-CFTR Mouse Monoclonal Antibody [clone: CF3]
Anticuerpos
The CFTR Antibody (CF3) from Novus Biologicals is a mouse monoclonal antibody to CFTR. This antibody reacts with human, mouse, rat, chicken. The CFTR Antibody (CF3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Blocking/Neutralising.

Type: Primary
Antigen: CFTR
Clonality: Monoclonal
Clone: CF3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR