Anti-HSPA5 Rabbit Polyclonal Antibody (DyLight® 405)

Proveedor: Novus Biologicals
NB300-520V
NOVUNB300-520VEA 710 EUR
NOVUNB300-520V
Anti-HSPA5 Rabbit Polyclonal Antibody (DyLight® 405)
Anticuerpos
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR