Anti-KDM6B Rabbit Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Novus Biologicals
NBP1-06640F
NOVUNBP1-06640FEA 768 EUR
NOVUNBP1-06640F
Anti-KDM6B Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: KDM6B (lysine (K)-specific demethylase 6B)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR