Anti-Palladin Mouse Monoclonal Antibody (DyLight® 650) [clone: 1E6]

Proveedor: Novus Biologicals
NBP1-25959C
NOVUNBP1-25959CEA 722 EUR
NOVUNBP1-25959C
Anti-Palladin Mouse Monoclonal Antibody (DyLight® 650) [clone: 1E6]
Anticuerpos
Type: Primary
Antigen: Palladin
Clonality: monoclonal
Clone: 1000000
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR