Anti-SCD Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Novus Biologicals
NBP1-26393H
NOVUNBP1-26393HEA 722 EUR
NOVUNBP1-26393H
Anti-SCD Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: SCD (stearoyl-CoA desaturase (delta-9-desaturase))
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR