Anti-DUX4 Mouse Monoclonal Antibody (DyLight® 488) [clone: P4H2]

Proveedor: Novus Biologicals
NBP1-49552G
NOVUNBP1-49552GEA 650 EUR
NOVUNBP1-49552G
Anti-DUX4 Mouse Monoclonal Antibody (DyLight® 488) [clone: P4H2]
Anticuerpos
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone: P4H2
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR