Anti-F2R Mouse Monoclonal Antibody (DyLight® 405) [clone: N2-11]

Proveedor: Novus Biologicals
NBP1-71770V
NOVUNBP1-71770VEA 722 EUR
NOVUNBP1-71770V
Anti-F2R Mouse Monoclonal Antibody (DyLight® 405) [clone: N2-11]
Anticuerpos
Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: monoclonal
Clone: N2-11
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR