Anti-FAT4 Rabbit Polyclonal Antibody (DyLight® 650)

Proveedor: Novus Biologicals
NBP1-78381C
NOVUNBP1-78381CEA 668 EUR
NOVUNBP1-78381C
Anti-FAT4 Rabbit Polyclonal Antibody (DyLight® 650)
Anticuerpos
Type: Primary
Antigen: FAT4 (FAT atypical cadherin 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR