Anti-ZNF521 Rabbit Polyclonal Antibody (DyLight® 550)

Proveedor: Novus Biologicals
NBP1-91270R
NOVUNBP1-91270REA 668 EUR
NOVUNBP1-91270R
Anti-ZNF521 Rabbit Polyclonal Antibody (DyLight® 550)
Anticuerpos
Type: Primary
Antigen: ZNF521 (zinc finger protein 521)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR