Anti-SUV420H1 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP1-97312 NBP1-97312SS
NOVUNBP1-97312EA 554 EUR
NOVUNBP1-97312 NOVUNBP1-97312SS
Anti-SUV420H1 Rabbit Polyclonal Antibody
Anticuerpos
The SUV420h1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SUV420h1. This antibody reacts with human, mouse. The SUV420h1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: SUV420h1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR