Anti-TRPV1 Rabbit Polyclonal Antibody (DyLight® 550)

Proveedor: Novus Biologicals
NBP1-97417R
NOVUNBP1-97417REA 679 EUR
NOVUNBP1-97417R
Anti-TRPV1 Rabbit Polyclonal Antibody (DyLight® 550)
Anticuerpos
Type: Primary
Antigen: TRPV1 (transient receptor potential cation channel, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR