Anti-RSAD2 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP2-03971
NOVUNBP2-03971EA 468 EUR
NOVUNBP2-03971
Anti-RSAD2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RSAD2 (radical S-adenosyl methionine domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR