Anti-RPS6 Goat Polyclonal Antibody

Proveedor: Novus Biologicals
NBP2-23649
NOVUNBP2-23649EA 444 EUR
NOVUNBP2-23649
Anti-RPS6 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RPS6 (ribosomal protein S6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR