Anti-LXN Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NBP2-23723
NOVUNBP2-23723EA 434 EUR
NOVUNBP2-23723
Anti-LXN Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LXN (latexin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR