Disposable syringes
Proveedor: THERMO NATIONAL SCIENTIFIC
S7515-10
NSCAS7515-10EA 271 EUR
NSCAS7515-10
Disposable syringes
Jeringas Jeringas para cromatografía
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
  • Non sterile
  • Bulk packed