Bood DNA HV kit, CE IVD, Mag-Bind®
Proveedor: OMEGA BIO-TEK
M3292-03CEIVD
OMEGM3292-03CEIVDEA 2720 EUR
OMEGM3292-03CEIVD
Bood DNA HV kit, CE IVD, Mag-Bind®
Reactivos de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples.
The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples. Mag-Bind® Particles CH provide a quick magnetic response time reducing overall processing time. This system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proven efficiency of Omega Bio-Tek’s DNA isolation system to provide a fast and convenient method to isolate DNA. Utilizing paramagnetic particles provides high-quality DNA that is suitable for direct use in most downstream applications, such as amplification and enzymatic reactions. The 4 ml whole blood protocol has been successfully automated on the Hamilton Microlab® STAR™.
Certificaciones: CE-IVD IVDR