Anti-FAM119A Mouse Monoclonal Antibody [clone: 2D6]

Proveedor: OriGene
CF503703
ORIGCF503703EA 833 EUR
ORIGCF503703
Anti-FAM119A Mouse Monoclonal Antibody [clone: 2D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FAM119A
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno FAM119A
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 2D6
Reactividad Human
Huésped Mouse
Isotipo IgG1
Western blot Yes
Epítopo Full length
Inmunógeno Full length human recombinant protein of human FAM119A(NP_660323) produced in HEK293T cell.
Forma Lyophilized
Molecular weight 24.4 kDa
Purification purified by affinity chromatography
Adsorción cruzada No
Storage buffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Temperatura de almacenamiento –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR