Anti-MDM4 Mouse Monoclonal Antibody [clone: 4D2]

Proveedor: OriGene
CF505727
ORIGCF505727EA 833 EUR
ORIGCF505727
Anti-MDM4 Mouse Monoclonal Antibody [clone: 4D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MDM4
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno MDM4
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 4D2
Reactividad Human
Huésped Mouse
Isotipo IgG2b
Western blot Yes
FluorescenciaInmunológica Yes
Epítopo Full length
Inmunógeno Full length human recombinant protein of human MDM4(NP_002384) produced in HEK293T cell.
Forma Lyophilized
Molecular weight 54.7 kDa
Purification purified by affinity chromatography
Adsorción cruzada No
Storage buffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Temperatura de almacenamiento –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR