Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 4F1]

Proveedor: OriGene
CF505767
ORIGCF505767EA 833 EUR
ORIGCF505767
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 4F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 4F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR