Anti-BOLL Mouse Monoclonal Antibody [clone: 2D11]

Proveedor: OriGene
CF804110
ORIGCF804110EA 833 EUR
ORIGCF804110
Anti-BOLL Mouse Monoclonal Antibody [clone: 2D11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: BOLL
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR