Anti-MMAB Mouse Monoclonal Antibody [clone: OTI2G5]

Proveedor: OriGene
TA502178
ORIGTA502178EA 621 EUR
ORIGTA502178
Anti-MMAB Mouse Monoclonal Antibody [clone: OTI2G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MMAB (methylmalonic aciduria (cobalamin deficiency) cblB type)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR