Anti-EMC8 Mouse Monoclonal Antibody [clone: OTI5H3]

Proveedor: OriGene
TA507067
ORIGTA507067EA 621 EUR
ORIGTA507067
Anti-EMC8 Mouse Monoclonal Antibody [clone: OTI5H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: EMC8 (ER membrane protein complex subunit 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno EMC8
Environmentally Preferable
Nombre del antígeno ER membrane Protein complex subunit 8
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon OTI5H3
Reactividad Human
Huésped Mouse
Isotipo IgG1
Western blot Yes
Epítopo Full length
Forma Liquid
Molecular weight 23.6 kDa
Purification purified by affinity chromatography
Adsorción cruzada none
Storage buffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR