Anti-KRT5 Mouse Monoclonal Antibody [clone: OTI6F8]

Proveedor: OriGene
TA800727
ORIGTA800727EA 621 EUR
ORIGTA800727
Anti-KRT5 Mouse Monoclonal Antibody [clone: OTI6F8]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR