Anti-CDKN2C Mouse Monoclonal Antibody [clone: OTI2E1]

Proveedor: OriGene
TA801184
ORIGTA801184EA 621 EUR
ORIGTA801184
Anti-CDKN2C Mouse Monoclonal Antibody [clone: OTI2E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CDKN2C (cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR