Orion Star™ T910 pH Titrators
Proveedor: Thermo Orion
START9101 START9100 START9102
ORIOSTART9101EA 4300 EUR
ORIOSTART9101 ORIOSTART9100 ORIOSTART9102
Orion Star™ T910 pH Titrators
Valoradores
The Thermo Scientific™ Orion Star™ T910 pH titrators are designed to increase your laboratory productivity by automating titrations. Our core electrochemistry technology is integrated with a state-of-the-art reagent dispensing system to create modern, simplified automated titrators designed to make performing titrations easier, more reliable and more reproducible than manual titrations.
  • Accurate, reproducible results–automating the titration workflow enables analysis to be run using the same setup parameters and calculations every time
  • Fast setup time–