rubis® Tweezers Set
Proveedor: OUTILS RUBIS
1K531
OUTI1K531EA 157 EUR
OUTI1K531
rubis® Tweezers Set
Pinzas
Set of 5 tweezers containing 1 tweezer 00-SA, 1 tweezer 2A-SA, 1 tweezer 3C-SA, 1 tweezer 5-SA, and 1 tweezer 7-SA.
  • Made from stainless steel
  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic
  • Packaged in a practical hardcover case
Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology, and electron microscopy.