Culti-Loops™ Staphylococcus epidermidis ATCC™ 700296™
Proveedor: THERMO FISHER DIAGNOSTICS
R4609457
OXOIR4609457EA 262.5 EUR
OXOIR4609457
Culti-Loops™ Staphylococcus epidermidis ATCC™ 700296™
Tests para microbiología Tests de microorganismos
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Envase: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.