Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Proveedor: THERMO FISHER DIAGNOSTICS
R4609498
OXOIR4609498EA 262.5 EUR
OXOIR4609498
Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Tests para microbiología Tests de microorganismos
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Culti-Loops™ QC organisms.
The loop may be dissolved in a liquid medium. Alternatively, it can be streaked directly onto appropriate media.
Envase: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.