Transfection boosters, GeneBlaster™
Proveedor: OZ BIOSCIENCES
GB20010 GB20011 GB20012 GB20013 GB20014
OZBIGB20010EA 162.95 EUR
OZBIGB20010 OZBIGB20011 OZBIGB20012 OZBIGB20013 OZBIGB20014
Transfection boosters, GeneBlaster™
Reactivos para transfección
GeneBlaster™ transfection boosters are an innovative and efficient solution to improving gene expression levels. GeneBlaster™ kits are a set of chemicals designed to get higher and longer transgene expression levels.
  • Higher gene expression in many cells
  • Convenient for a large panel of adherent and suspension cells
  • Prolong in vitro gene expression
  • Successful with all genetic vectors
  • Simple, rapid, and easy to use
Store all GeneBlaster™ kits at +4 °C, except GeneBlaster™ Emerald at –20 °C.
Información de suministro: Shipped at room temperature.
Order Now
ESPECIFICACIONES
Celda específica Hematopoietic suspension Neurons