Perileno ≥99.0%, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

P11204-1G P11204-5G
P11204-1GEA 46.8 EUR
P11204-1G P11204-5G
Perileno ≥99.0%, Sigma-Aldrich®
Perileno

The polycyclic aromatic hydrocarbon perylene is a hydrophobic fluorescent probe. It is highly photostable and displays blue fluorescence. Perylene is used as a dye for spectroscopic studies.


Perylene has been used as a standard along with a mixture of polycyclic aromatic hydrocarbons (PAHs) to determine the spatial distribution and sources of PAHs in surface sediments of rivers.

Formula: C₂₀H₁₂
Pto. Ebullición: 443 °C (1013 hPa)
Pto. de fusión: 276…279 °C (lit.)
Densidad: 1,362 g/cm³ (20 °C)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00004142
Núm. CAS: 198-55-0
EINECS: 205-900-9
Índice Merck: 12,07325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fecd171289d227be926b874d4a5001d67e77fe55&vid=6ff19028cb9ccd06fd2f57b58d5a140093f9ced8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Gold to orange crystalline powder
Infrared spectrum Conforms to structure.
Elemental analysis Carbon 94.2% - 96.2%
High pressure liquid 99.0% (minimum)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR