Ponceau S colorante, bioreagent Electrophoresis, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

MQ200

P3504-100G P3504-10G P3504-50G
P3504-100GEA 299 EUR
P3504-100G P3504-10G P3504-50G
Ponceau S colorante, bioreagent Electrophoresis, Sigma-Aldrich®
3-Hidroxi-4-(2-sulfonato-4-(4-sulfonatofenilazo)fenilazo)naftaleno-2,7-disulfonato de tetrasodio

Ponceau S is the sodium salt of a diazo dye. It is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. The stain is reversed with water washes and aids in subsequent immunological detection. The following are two common stain formulations: 0.1% Ponceau S (w/v) in 5% acetic acid 2% Ponceau S (w/v) in 30% TCA, 30% sulfosalicylic acid.


  • Ponceau S has been used for membrane staining (Western blotting)

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00003892
Núm. CAS: 6226-79-5
EINECS: 228-319-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00732e6c5db3fa4d9811900b38dc089c2eee2b41&vid=ae83338aee7cdb702d714e8689bc41725c033bde&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Infrared spectrum Conforms to Structure
Solubility (Turbidity) Clear
Solubility (Colour)
c = 4mg/4ml; Water
Red to Very Dark Red
Suitability
Suitable for Use as a Stain in Electrophoresis
Suitable
Wavelength
C = 0.015 G/L, Di water
518 - 522 nm
Extinction Coefficient ≥33000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR