Pararosaniline, crystalline, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base

M-Clarity™ quality level = MQ200

P7632-10G P7632-1G P7632-25G P7632-5G
P7632-10GEA 69.5 EUR
P7632-10G P7632-1G P7632-25G P7632-5G
Pararosaniline, crystalline, Sigma-Aldrich®
Pararosaniline

Pararosaniline Base,crystalline (Parafuchsin; Paramagenta)

Formula: C₁₉H₁₉N₃O
Pto. de fusión: 205 °C (desc., lit.)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00036222
Núm. CAS: 467-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e36cdecf2bf49839b29f2c1236505faa634fe850&vid=99fb0993b82e68600ae7cd3240adda90479d65a7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Colour) Light Red to Very Dark Purple
Appearance (Form) Powder
Solubility (Colour) Lt. Pink to Red to Very Dark Purple
Solubility (Turbidity)
1 mg/ml, EtOH
Clear to Hazy
Infrared spectrum Conforms
Carbon 70.2 - 79.2 %
Nitrogen 12.9 - 14.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR