Utility carts
Proveedor: Palbam
UC-85-2 UC-85-3 UCP-85-2 UC-55-2 UC-55-3 UCP-55-3 UCP-85-3 UCP-55-2 UC-85-2-ESD UCP-55-3-ESD
139-0589EA 2300 EUR
139-0589 139-0590 139-0591 139-0588 139-0793 139-0794 139-0795 PALBUCP-55-2 PALBUC-85-2-ESD PALBUCP-55-3-ESD
Utility carts
Carritos
These cleanroom carts are designed to withstand heavy loads.
  • Up to 60 kg per shelf
  • Integral raised ledges prevent parts from rolling off and are easy to wipe down
  • Choice of two or three shelves
  • Solid or perforated
  • White nylon 130 mm (5") cleanroom castors (Two with brakes) supplied with every cart
Stainless steel 304 with electropolished finish. Special non-marking, smooth running.
Información de suministro: Typically available from stock for direct shipment from the supplier. Shipped flat-packed with assembly instructions.