Membrane nitrogen generator, Infinity XE 60 series
Proveedor: PEAK SCIENTIFIC
3303147
PEAK3303147EA 13560 EUR
PEAK3303147
Membrane nitrogen generator, Infinity XE 60 series
Generadores de gas Generadores de nitrógeno
Producing between 10 and 520 L/min of high purity nitrogen gas, the Infinity XE 60 series nitrogen membrane gas generators can comfortably supply multiple laboratory instruments with instrument grade nitrogen.
  • Flow rates ranging from 10 to 520 L/min *
  • N₂ purity up to 99,5%
  • Versatile solution with varying nitrogen flow, pressure and purity capabilities
  • In-field expansion of nitrogen capacity, for models without dry air outlet, to meet growing laboratory demands
  • Optional second outlet for dry air
  • Intelligent membrane management (IMM) optimises compressed air