SPE columns, Supra-Poly® HLB
Proveedor: PerkinElmer
N9306678 N9306679 N9306689 N9306650 N9306662 N9306661 N9306660 N9306659 N9306672 N9306666 N9306688 N9306674 N9306675 N9306676 N9306677 N9306680 N9306681 N9306682 N9306683 N9306684 N9306685 N9306687 N9306671 N9306690 N9306692 N9306693 N9306694 N9306686 N9306658 N9306651 N9306652 N9306653 N9306655 N9306673 N9306664 N9306670 N9306669 N9306668 N9306667 N9306656 N9306665 N9306657
PERCN9306678EA 311 EUR
PERCN9306678 PERCN9306679 PERCN9306689 PERCN9306650 PERCN9306662 PERCN9306661 PERCN9306660 PERCN9306659 PERCN9306672 PERCN9306666 PERCN9306688 PERCN9306674 PERCN9306675 PERCN9306676 PERCN9306677 PERCN9306680 PERCN9306681 PERCN9306682 PERCN9306683 PERCN9306684 PERCN9306685 PERCN9306687 PERCN9306671 PERCN9306690 PERCN9306692 PERCN9306693 PERCN9306694 PERCN9306686 PERCN9306658 PERCN9306651 PERCN9306652 PERCN9306653 PERCN9306655 PERCN9306673 PERCN9306664 PERCN9306670 PERCN9306669 PERCN9306668 PERCN9306667 PERCN9306656 PERCN9306665 PERCN9306657
SPE columns, Supra-Poly® HLB
Columnas de cromatografía
  • Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
  • Shorter analysis times, greater load capacity and reduced solvent usage
  • Ideal for high throughput assays