Capillary columns, Elite-5 for autosystem
Proveedor: PerkinElmer
N9316276 N9316066 N9316067 N9316068 N9316075 N9316083 N9316092 N9316099 N9316272 N9316274 N9316287 N9316289 N9316290 N9316303 N9316069 N9316073 N9316076 N9316077 N9316079 N9316082 N9316086 N9316087 N9316088 N9316090 N9316091 N9316098 N9316100 N9316102 N9316103 N9316110 N9316111 N9316112 N9316273 N9316275 N9316279 N9316282 N9316283 N9316284 N9316286 N9316293 N9316294 N9316295 N9316297 N9316298 N9316300 N9316302
PERCN9316276EA 1010 EUR
PERCN9316276 PERCN9316066 PERCN9316067 PERCN9316068 PERCN9316075 PERCN9316083 PERCN9316092 PERCN9316099 PERCN9316272 PERCN9316274 PERCN9316287 PERCN9316289 PERCN9316290 PERCN9316303 PERCN9316069 PERCN9316073 PERCN9316076 PERCN9316077 PERCN9316079 PERCN9316082 PERCN9316086 PERCN9316087 PERCN9316088 PERCN9316090 PERCN9316091 PERCN9316098 PERCN9316100 PERCN9316102 PERCN9316103 PERCN9316110 PERCN9316111 PERCN9316112 PERCN9316273 PERCN9316275 PERCN9316279 PERCN9316282 PERCN9316283 PERCN9316284 PERCN9316286 PERCN9316293 PERCN9316294 PERCN9316295 PERCN9316297 PERCN9316298 PERCN9316300 PERCN9316302
Capillary columns, Elite-5 for autosystem
Columnas de cromatografía
Elite-5 and Elite-5ht columns are suitable for semivolatiles, phenols, amines, residual solvents, drugs of abuse, pesticides, PCB congeners (e.g. Aroclor mixes) and solvent impurities.
Elite-5MS capillary columns are general-purpose columns – ideal for GCMS analysis of semivolatiles, polycyclic aromatic compounds, chlorinated hydrocarbons, phthalates, phenols, amines, organochlorine pesticides, organophosphorus pesticides, drugs, solvent impurities, and hydrocarbons.