Capillary columns, Elite-Volatiles for autosystem
Proveedor: PerkinElmer
N9316388 N9316390 N9316391 N9316393
PERCN9316388EA 4350 EUR
PERCN9316388 PERCN9316390 PERCN9316391 PERCN9316393
Capillary columns, Elite-Volatiles for autosystem
Columnas de cromatografía
Elite Volatiles specialty phase capillary columns are ideal for the analysis of volatile organic compounds.