Push brooms, soft sweep
Proveedor: PERFEX
2624B
PERF2624BEA 54.3 EUR
PERF2624B
Push brooms, soft sweep
Escobas
These brooms are made of extremely durable polypropylene fibres which are non-conductive and will not absorb bacteria, liquids, or odours.
This specially formulated polypropylene fill is excellent for sweeping very fine and medium particles from resilient smooth floors, vinyl, linoleum, terrazzo, marble, tile, finished wood and smooth concrete.
Información para pedidos: Polymer adjustable or fixed length handles must be ordered separately.