Iodination beads, Pierce™
Proveedor: Thermo Fisher Scientific
28665
PIER28665EA 233 EUR
PIER28665
Iodination beads, Pierce™
Perlas Bolas de plástico
Pierce™ Iodination beads are Ø 3 mm polystyrene beads that are coated with an oxidising reagent, which provides efficient activation of 125I for protein or peptide iodination procedures.
  • Derivatised, uniform, non porous polystyrene beads
  • Iodine-125 incorporation as high as 99%; labeled protein recovery >90%
  • Able to function over a broad pH and temperature range
  • Iodinates in the presence of azides, detergents, urea and high salt
  • Allows efficient iodination of cell membrane surface proteins
  • Fast and easy to use – iodinations complete in 2 to 15 minutes