Horseradish Peroxidase, Biotinylated, Pierce™
Proveedor: Thermo Fisher Scientific
29139
PIER29139EA 216 EUR
PIER29139
Horseradish Peroxidase, Biotinylated, Pierce™
Ensayos Ensayos con proteinas
Biotinylated horse radish peroxidase is used as control or signal amplification in IHC to amplify the signal of biotinylated primary antibodies using the avidin-biotin complex staining method.