Pierce™ Recombinant Protein G, Biotinylated
Proveedor: Thermo Fisher Scientific
29988
PIER29988EA 274 EUR
PIER29988
Pierce™ Recombinant Protein G, Biotinylated
Sistemas para purificación de proteínas Reactivos para purificación de proteinas
Biotinylated recombinant Protein G can be used to probe and detect mouse and human antibodies, especially IgG isotypes, in western blotting, ELISA, IHC and other immunoassay protocols.
  • Contains two Fc-binding domains per protein
  • Does not bind human IgM, IgD or IgA
  • Efficient than Protein A for mouse (IgG1), human (IgG3) and rat, goat and cow antibodies
  • Less efficient than Protein A for guinea pig, pig, dog and cat