Pierce™ Peroxidase Suppressor
Proveedor: Thermo Fisher Scientific
35000
PIER35000EA 191 EUR
PIER35000
Pierce™ Peroxidase Suppressor
Ensayos Ensayos inmunológicos
This peroxidase suppressor is a stable and easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues.
  • Stable and ready-to-use
  • Effectively inhibits peroxidase activity
  • No interference with antigen-antibody interactions
Designed to reduce background signal in HRP-based immunohistochemistry applications.
Precaución: Store at 4 °C upon receipt.