Dialysis tubing, SnakeSkin™, Pierce™
Proveedor: Thermo Fisher Scientific
68100 68035 68700 88242 88243 88244 88245
PIER68100EA 247 EUR
PIER68100 PIER68035 PIER68700 PIER88242 PIER88243 PIER88244 PIER88245
Dialysis tubing, SnakeSkin™, Pierce™
Tuberías Tuberías para diálisis
SnakeSkin dialysis tubings with pleated format are made from high performance regenerated-cellulose membrane which has minimal nonspecific binding and a reliable molecular-weight cutoff (MWCO).
  • Ready-to-use, pre-opened and pre-wetted
  • Greater than 90% protein recovery
Regenerated cellulose is exceptionally stable when exposed to common laboratory solutions such as acidic and basic conditions, or hydrophobic solvents and alcohols. Ideal for dialysing protein samples.
Información de suministro: Supplied in eight-inch (20 cm) cylinders containing 35More Product Information