Pierce™ Protein Concentrators
Proveedor: Thermo Fisher Scientific
88512 88513 88502 88503 88527 88516 88521 88522 88523 88524 88525 88526 88528 88529 88531 88532 88533 88534 88535 88536 88537 88514 88515 88517
PIER88512EA 153 EUR
PIER88512 PIER88513 PIER88502 PIER88503 PIER88527 PIER88516 PIER88521 PIER88522 PIER88523 PIER88524 PIER88525 PIER88526 PIER88528 PIER88529 PIER88531 PIER88532 PIER88533 PIER88534 PIER88535 PIER88536 PIER88537 PIER88514 PIER88515 PIER88517
Pierce™ Protein Concentrators
Filtros Filtros centrífugos
Protein Concentrators PES are disposable ultrafiltration centrifugal devices with a polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples such as proteins and nucleic acids.
  • Sample concentration of 10- to 30-fold can be achieved in 10 minutes or less
  • Unique design minimises membrane fouling and provides high speed concentrations, even with particle laden solutions
  • Compatible with most bench-top microcentrifuges with fixed-angle rotors that accommodate 2,2 ml tubes
  • Molecular weight cutoff
  • Can be used for