Protein A/G magnetic beads, Pierce™
Proveedor: Thermo Fisher Scientific
88802 88803
PIER88802EA 257 EUR
PIER88802 PIER88803
Protein A/G magnetic beads, Pierce™
Perlas Bolas magnéticas
Pierce™ protein A/G magnetic beads are high-performance affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.
  • Stable, pre-blocked beads provide clean purification products
  • Flexible for IgG binding domains of both Protein A and Protein G on one bead
  • Beads are compatible with manual and automated applications
Eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes. Coated with genetically engineeredMore Product Information