Transfection reagent, Turbofect™
Proveedor: Thermo Fisher Scientific
R0533 R0534
PIERR0533EA 71.75 EUR
PIERR0533 PIERR0534
Transfection reagent, Turbofect™
Reactivos para transfección
TurboFect™ reagent an easy to use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
  • High transfection efficiency of a wide variety of cell types
  • No optimisation required for most cell lines
  • Excellent transfection efficiency in the presence or absence of serum
  • Minimal cytotoxicity
  • Ready to use without any need to reconstitute, dilute or manipulate
TurboFect™ reagent is ideal for stable and transient transfection or co-transfection of DNA or RNA, experiments with adherent or suspension cells, differentiated or undifferentiated cells and gene expression studies and assays involving GFP or luciferase reporters.