Scandium standard solution, for ICP, in HNO₃, CRM, VHG
Proveedor: LGC Standards PROMOCHEM
VHG-PSCN-100 VHG-LSCN-100 VHG-PSCN-500 VHG-TSCN-100 VHG-TSCN-500
PROOVHG-PSCN-100EA 100 EUR
PROOVHG-PSCN-100 PROOVHG-LSCN-100 PROOVHG-PSCN-500 PROOVHG-TSCN-100 PROOVHG-TSCN-500
Scandium standard solution, for ICP, in HNO₃, CRM, VHG
Normativas Estándares catiónicos Estándares de escandio
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaciones: Traceable to NIST, SRM.